Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR4A2
Entrez Gene ID
697077
Gene Name
nuclear receptor subfamily 4 group A member 2
Gene Aliases
EGK_04449
Chromosome Location
Chr.12: 43420121 - 43428427 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266910.2 NP_001253839.1 - 212 74
XM_015110132.2 XP_014965618.1 - 414 74
XM_015110133.2 XP_014965619.1 - 414 74
GenBank mRNA JU471922.1 - - 212 74
JU471923.1 - - 212 74

View Full Product Details