Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC1
Entrez Gene ID
697969
Gene Name
zinc finger DHHC-type containing 1
Gene Aliases
-
Chromosome Location
Chr.20: 54580497 - 54603129 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126393.2 XP_014981879.1 - 805 61
XM_015126394.2 XP_014981880.1 - 915 61
XM_015126397.2 XP_014981883.1 - 807 61
XM_015126399.2 XP_014981885.1 - 303 61
XM_028840934.1 XP_028696767.1 - 1417 61
XM_028840935.1 XP_028696768.1 - 886 61

View Full Product Details