Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC6A11
Entrez Gene ID
696741
Gene Name
solute carrier family 6 member 11
Gene Aliases
GAT3
Chromosome Location
Chr.2: 142228861 - 142351426 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134952.2 NP_001128424.1 - 455 81
GenBank mRNA JU327115.1 - - 270 81
JV529211.1 - - 429 81
JV564962.1 - - 427 81

View Full Product Details