Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUCO
Entrez Gene ID
705776
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1H1orf9
Chromosome Location
Chr.1: 29836904 - 29918532 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001100732.4 XP_001100732.2 - 3644 150
XM_015124042.2 XP_014979528.2 - 3365 150
XM_028850415.1 XP_028706248.1 - 3490 150
GenBank mRNA JU334889.1 - - 3595 150
JU334890.1 - - 3595 150
JU334891.1 - - 3595 150
JU475899.1 - - 3542 150
JV047300.1 - - 3601 150
JV047301.1 - - 3580 150
JV636208.1 - - 3558 150

View Full Product Details