Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRINP1
Entrez Gene ID
701794
Gene Name
BMP/retinoic acid inducible neural specific 1
Gene Aliases
DBC1, EGK_07379
Chromosome Location
Chr.15: 23366085 - 23564613 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260978.1 NP_001247907.1 - 802 69
GenBank mRNA JU320643.1 - - 802 69
JV565994.1 - - 873 69

View Full Product Details