Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPGS2
Entrez Gene ID
709953
Gene Name
tubulin polyglutamylase complex subunit 2
Gene Aliases
C18orf10, EGK_09642
Chromosome Location
Chr.18: 45726560 - 45775006 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260587.2 NP_001247516.1 - 417 127
XM_015121729.2 XP_014977215.1 - 376 127
XM_015121733.2 XP_014977219.1 - 607 127
XM_015121734.2 XP_014977220.1 - 607 127
XM_015121735.2 XP_014977221.1 - 377 127
GenBank mRNA DQ148062.1 - - 162 127
JU321611.1 - - 364 127
JU475069.1 - - 416 127
JV046690.1 - - 418 127
JV612369.1 - - 431 127

View Full Product Details