Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPGS2
Entrez Gene ID
709953
Gene Name
tubulin polyglutamylase complex subunit 2
Gene Aliases
C18orf10, EGK_09642
Chromosome Location
Chr.18: 45726560 - 45775006 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260587.2 NP_001247516.1 - 636 108
XM_015121730.2 XP_014977216.1 - 696 108
XM_015121731.2 XP_014977217.1 - 573 108
XM_015121733.2 XP_014977219.1 - 826 108
XM_015121734.2 XP_014977220.1 - 826 108
XM_015121735.2 XP_014977221.1 - 596 108
GenBank mRNA DQ148062.1 - - 381 108
JU321611.1 - - 583 108
JU475069.1 - - 635 108
JV046690.1 - - 637 108
JV612369.1 - - 650 108

View Full Product Details