Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP4
Entrez Gene ID
701543
Gene Name
THAP domain containing 4
Gene Aliases
-
Chromosome Location
Chr.12: 129025003 - 129070854 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015111564.2 XP_014967050.1 - 1539 110
GenBank mRNA JU334773.1 - - 1294 110
JU475814.1 - - 1269 110
JV047240.1 - - 1282 110

View Full Product Details