Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNRK
Entrez Gene ID
715803
Gene Name
SNF related kinase
Gene Aliases
EGK_11746
Chromosome Location
Chr.2: 99228890 - 99292258 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257661.1 NP_001244590.1 - 1227 112
XM_015131175.1 XP_014986661.1 - 1326 112
GenBank mRNA JU322234.1 - - 1227 112
JU476134.1 - - 1141 112
JV717642.1 - - 1221 112

View Full Product Details