Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THG1L
Entrez Gene ID
715853
Gene Name
tRNA-histidine guanylyltransferase 1 like
Gene Aliases
-
Chromosome Location
Chr.6: 155150492 - 155160536 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194604.2 NP_001181533.2 - 310 136
GenBank mRNA JU476165.1 - - 214 136

View Full Product Details