Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBCCD1
Entrez Gene ID
700153
Gene Name
TBCC domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 91884435 - 91905000 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266952.1 NP_001253881.1 - 599 80
XM_015131361.2 XP_014986847.1 - 1194 80
GenBank mRNA JU335249.1 - - 595 80
JU472675.1 - - 599 80

View Full Product Details