Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM19
Entrez Gene ID
718700
Gene Name
transmembrane protein 19
Gene Aliases
EGK_03932
Chromosome Location
Chr.11: 71235018 - 71250372 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261114.1 NP_001248043.1 - 1069 117
GenBank mRNA JU322717.1 - - 917 117
JU470866.1 - - 1190 117
JV045191.1 - - 1069 117
JV637271.1 - - 1209 117

View Full Product Details