Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT4
Entrez Gene ID
715385
Gene Name
nudix hydrolase 4
Gene Aliases
-
Chromosome Location
Chr.11: 93181170 - 93205756 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261255.1 NP_001248184.1 - 659 144
XM_015152398.2 XP_015007884.1 - 390 144
GenBank mRNA JU322193.1 - - 645 144
JU471425.1 - - 659 144

View Full Product Details