Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADPRM
Entrez Gene ID
717991
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C16H17orf48, C17orf48
Chromosome Location
Chr.16: 10487251 - 10508305 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194621.1 NP_001181550.1 - 640 89
XM_015118508.2 XP_014973994.1 - 725 89
XM_015118509.2 XP_014973995.1 - 725 89
XM_015118510.2 XP_014973996.1 - 726 89
GenBank mRNA JU335547.1 - - 643 89
JU476474.1 - - 690 89
JV047712.1 - - 682 89
JV656459.1 - - 597 89

View Full Product Details