Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL7
Entrez Gene ID
714204
Gene Name
angiopoietin like 7
Gene Aliases
-
Chromosome Location
Chr.1: 213263477 - 213270203 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001103825.4 XP_001103825.1 - 761 119
GenBank mRNA JU478642.1 - - 582 119

View Full Product Details