Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP26
Entrez Gene ID
574248
Gene Name
matrix metallopeptidase 26
Gene Aliases
EGK_04430
Chromosome Location
Chr.14: 63516469 - 63520705 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115062.1 XP_014970548.1 - 339 118
GenBank mRNA AY090610.1 - - 137 118

View Full Product Details