Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GID4
Entrez Gene ID
699842
Gene Name
GID complex subunit 4 homolog
Gene Aliases
C16H17orf39, C17orf39
Chromosome Location
Chr.16: 17875045 - 17905887 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193962.2 NP_001180891.1 - 766 121

View Full Product Details