Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H1orf50
Entrez Gene ID
698058
Gene Name
chromosome 1 C1orf50 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 181781136 - 181790257 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266364.1 NP_001253293.1 - 234 116
GenBank mRNA JU323221.1 - - 215 116
JU477169.1 - - 234 116
JV593347.1 - - 249 116

View Full Product Details