Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FA2H
Entrez Gene ID
710403
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
-
Chromosome Location
Chr.20: 61676099 - 61739583 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194422.3 NP_001181351.2 - 584 115
GenBank mRNA JU336356.1 - - 565 115
JV723233.1 - - 491 115

View Full Product Details