Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
713876
Gene Name
FA core complex associated protein 100
Gene Aliases
C16H17orf70, C17orf70
Chromosome Location
Chr.16: 77878390 - 77891037 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194508.2 NP_001181437.1 - 240 115
XM_015120543.2 XP_014976029.1 - 235 115
XM_015120544.2 XP_014976030.1 - 235 115
GenBank mRNA JU477304.1 - - 238 115
JV662181.1 - - 161 115

View Full Product Details