Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHD1
Entrez Gene ID
717627
Gene Name
EF-hand domain family member D1
Gene Aliases
-
Chromosome Location
Chr.12: 120058939 - 120110397 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257942.1 NP_001244871.1 - 484 81
GenBank mRNA JU336498.1 - - 484 81
JV521364.1 - - 118 81

View Full Product Details