Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REEP4
Entrez Gene ID
707267
Gene Name
receptor accessory protein 4
Gene Aliases
EGK_18750
Chromosome Location
Chr.8: 22302679 - 22306393 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266282.1 NP_001253211.1 - 1045 75
GenBank mRNA JU477316.1 - - 1141 75
JV048313.1 - - 1140 75
JV636389.1 - - 1045 75

View Full Product Details