Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRCIN1
Entrez Gene ID
693662
Gene Name
SRC kinase signaling inhibitor 1
Gene Aliases
SNIP
Chromosome Location
Chr.16: 49584573 - 49662640 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082491.4 XP_001082491.4 - 2083 134
XM_015119493.2 XP_014974979.2 - 2247 134
XM_015119494.2 XP_014974980.1 - 3671 134
XM_015119500.2 XP_014974986.1 - 1781 134
XM_028836428.1 XP_028692261.1 - 2049 134
XM_028836429.1 XP_028692262.1 - 2257 134
XM_028836430.1 XP_028692263.1 - 2246 134
XM_028836431.1 XP_028692264.1 - 2144 134
GenBank mRNA JU348927.1 - - 1735 134

View Full Product Details