Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
URM1
Entrez Gene ID
722618
Gene Name
ubiquitin related modifier 1
Gene Aliases
-
Chromosome Location
Chr.15: 12191174 - 12216032 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266132.1 NP_001253061.1 - 210 75
XM_015116635.2 XP_014972121.1 - 282 75
GenBank mRNA JU336544.1 - - 232 75
JU471574.1 - - 219 75
JU477360.1 - - 219 75
JV048340.1 - - 210 75
JV559251.1 - - 185 75

View Full Product Details