Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDCA3
Entrez Gene ID
722088
Gene Name
cell division cycle associated 3
Gene Aliases
-
Chromosome Location
Chr.11: 7014516 - 7017625 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194737.2 NP_001181666.1 - 473 78
XM_015150831.2 XP_015006317.1 - 875 78
XM_015150832.2 XP_015006318.1 - 875 78
GenBank mRNA JU473506.1 - - 157 78

View Full Product Details