Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R35
Entrez Gene ID
712067
Gene Name
protein phosphatase 1 regulatory subunit 35
Gene Aliases
-
Chromosome Location
Chr.3: 42507656 - 42509232 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104600.4 XP_001104600.2 - 908 114
XM_015134459.2 XP_014989945.2 - 933 114
GenBank mRNA JU325656.1 - - 217 114
JU482549.1 - - 213 114

View Full Product Details