Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAS1R2
Entrez Gene ID
714666
Gene Name
taste 1 receptor member 2
Gene Aliases
-
Chromosome Location
Chr.1: 205874414 - 205896125 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128089.1 NP_001121561.1 - 479 84

View Full Product Details