Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLHC1
Entrez Gene ID
716959
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
-
Chromosome Location
Chr.13: 53408280 - 53475621 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112371.2 XP_014967857.1 - 1599 112
XM_015112372.2 XP_014967858.1 - 1004 112
XM_028831672.1 XP_028687505.1 - 2050 112
XM_028831673.1 XP_028687506.1 - 829 112
XM_028831674.1 XP_028687507.1 - 514 112
XM_028831675.1 XP_028687508.1 - 284 112

View Full Product Details