Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH12
Entrez Gene ID
711438
Gene Name
retinol dehydrogenase 12
Gene Aliases
EGK_18313
Chromosome Location
Chr.7: 129841033 - 129874698 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194401.2 NP_001181330.1 - 394 123
XM_015143930.2 XP_014999416.1 - 454 123
GenBank mRNA JV457668.1 - - 206 123

View Full Product Details