Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM42
Entrez Gene ID
715465
Gene Name
tripartite motif containing 42
Gene Aliases
EGK_12049
Chromosome Location
Chr.2: 42838933 - 42861414 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113172.4 XP_001113172.3 - 2275 85

View Full Product Details