Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A4
Entrez Gene ID
705961
Gene Name
solute carrier family 10 member 4
Gene Aliases
-
Chromosome Location
Chr.5: 47898268 - 47904097 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001103529.4 XP_001103529.1 - 1059 93
GenBank mRNA JU340114.1 - - 411 93
JV709084.1 - - 491 93

View Full Product Details