Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEIOB
Entrez Gene ID
703322
Gene Name
meiosis specific with OB-fold
Gene Aliases
C16orf73, C20H16orf73
Chromosome Location
Chr.20: 1860079 - 1905590 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193903.2 NP_001180832.1 - 818 93
XM_028840457.1 XP_028696290.1 - 817 93
XM_028840458.1 XP_028696291.1 - 739 93
XM_028840459.1 XP_028696292.1 - 817 93
XM_028840460.1 XP_028696293.1 - 817 93
XM_028840461.1 XP_028696294.1 - 817 93
XM_028840462.1 XP_028696295.1 - 817 93
XM_028840463.1 XP_028696296.1 - 817 93
XM_028840464.1 XP_028696297.1 - 817 93
XM_028840465.1 XP_028696298.1 - 817 93
GenBank mRNA JV647373.1 - - 705 93

View Full Product Details