Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTRA4
Entrez Gene ID
700875
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
Chromosome Location
Chr.8: 39453340 - 39469007 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145209.2 XP_015000695.2 - 1071 76

View Full Product Details