Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LVRN
Entrez Gene ID
696886
Gene Name
laeverin
Gene Aliases
AQPEP, EGK_16767
Chromosome Location
Chr.6: 113548798 - 113623792 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086413.4 XP_001086413.1 - 3688 95

View Full Product Details