Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAAH2
Entrez Gene ID
705856
Gene Name
fatty acid amide hydrolase 2
Gene Aliases
EGK_20565
Chromosome Location
Chr.X: 56967966 - 57236632 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095907.4 XP_001095907.1 - 435 116
XM_015127556.2 XP_014983042.1 - 435 116

View Full Product Details