Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOP1
Entrez Gene ID
712780
Gene Name
otopetrin 1
Gene Aliases
-
Chromosome Location
Chr.5: 8763163 - 8800547 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097009.4 XP_001097009.2 - 426 160

View Full Product Details