Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCN12
Entrez Gene ID
708099
Gene Name
lipocalin 12
Gene Aliases
-
Chromosome Location
Chr.15: 1257502 - 1264258 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193934.3 NP_001180863.3 - 176 82
XM_015116303.2 XP_014971789.2 - 699 82
XM_015116304.2 XP_014971790.2 - 699 82
XM_015116305.2 XP_014971791.2 - 699 82
XM_028834313.1 XP_028690146.1 - 2018 82
XM_028834314.1 XP_028690147.1 - 699 82

View Full Product Details