Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19H19orf47
Entrez Gene ID
701191
Gene Name
chromosome 19 C19orf47 homolog
Gene Aliases
-
Chromosome Location
Chr.19: 39526156 - 39555142 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095658.4 XP_001095658.2 - 224 135
XM_001095873.4 XP_001095873.1 - 288 135
XM_015123974.2 XP_014979460.1 - 216 135
XM_015123975.2 XP_014979461.1 - 216 135
XM_015123976.2 XP_014979462.1 - 243 135
XM_015123977.2 XP_014979463.1 - 298 135
XM_015123978.2 XP_014979464.1 - 230 135
XM_015123979.2 XP_014979465.1 - 219 135
XM_028840192.1 XP_028696025.1 - 216 135
XM_028840193.1 XP_028696026.1 - 216 135
XM_028840194.1 XP_028696027.1 - 462 135
XM_028840195.1 XP_028696028.1 - 243 135
XM_028840196.1 XP_028696029.1 - 243 135
XM_028840197.1 XP_028696030.1 - 282 135
XM_028840198.1 XP_028696031.1 - 122 135
XM_028840199.1 XP_028696032.1 - 243 135
GenBank mRNA JU332154.1 - - 252 135
JU473803.1 - - 89 135
JV045718.1 - - 135 135

View Full Product Details