Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT26
Entrez Gene ID
702040
Gene Name
keratin 26
Gene Aliases
-
Chromosome Location
Chr.16: 51920911 - 51927296 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028835789.1 XP_028691622.1 - 1771 148

View Full Product Details