Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GKN2
Entrez Gene ID
700323
Gene Name
gastrokine 2
Gene Aliases
EGK_05400
Chromosome Location
Chr.13: 39847932 - 39855376 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028832130.1 XP_028687963.1 - 313 163

View Full Product Details