Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPS19BP1
Entrez Gene ID
704079
Gene Name
ribosomal protein S19 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 12012055 - 12015687 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JV700997.1 - - 134 118

View Full Product Details