Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSBN1L
Entrez Gene ID
693814
Gene Name
round spermatid basic protein 1 like
Gene Aliases
-
Chromosome Location
Chr.3: 128448709 - 128544631 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257886.1 NP_001244815.1 - 1341 86
GenBank mRNA JU333418.1 - - 1341 86
JU482565.1 - - 1332 86

View Full Product Details