Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMSAP2
Entrez Gene ID
709092
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1, EGK_05249
Chromosome Location
Chr.1: 58172263 - 58289558 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266713.1 NP_001253642.1 - 491 122
XM_015118795.2 XP_014974281.1 - 666 122
XM_015118805.2 XP_014974291.1 - 426 122
XM_015118811.2 XP_014974297.1 - 666 122
GenBank mRNA JU322212.1 - - 590 122
JV489114.1 - - 491 122
JV677854.1 - - 668 122

View Full Product Details