Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A10
Entrez Gene ID
696104
Gene Name
membrane spanning 4-domains A10
Gene Aliases
-
Chromosome Location
Chr.14: 13412347 - 13426939 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085216.4 XP_001085216.3 - 409 100
XM_015114091.2 XP_014969577.2 - 301 100
XM_028833379.1 XP_028689212.1 - 302 100
XM_028833380.1 XP_028689213.1 - 409 100

View Full Product Details