Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD12
Entrez Gene ID
704722
Gene Name
sterile alpha motif domain containing 12
Gene Aliases
-
Chromosome Location
Chr.8: 118454100 - 118927923 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145909.2 XP_015001395.1 - 360 98
XM_028852880.1 XP_028708713.1 - 422 98
XM_028852881.1 XP_028708714.1 - 463 98
XM_028852882.1 XP_028708715.1 - 409 98
GenBank mRNA JU334491.1 - - 319 98
JV529888.1 - - 203 98

View Full Product Details