Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDX1
Entrez Gene ID
710936
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
IPF1
Chromosome Location
Chr.17: 7144813 - 7150716 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001096758.4 XP_001096758.2 - 519 71

View Full Product Details