Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR52M1
Entrez Gene ID
717723
Gene Name
olfactory receptor, family 52, subfamily M, member 1
Gene Aliases
-
Chromosome Location
Chr.14: 64064917 - 64065870 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194566.2 NP_001181495.1 - 547 115

View Full Product Details