Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR51G2
Entrez Gene ID
716960
Gene Name
olfactory receptor, family 51, subfamily G, member 2
Gene Aliases
-
Chromosome Location
Chr.14: 63597759 - 63598784 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194547.1 NP_001181476.1 - 598 113

View Full Product Details