Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR10G2
Entrez Gene ID
708692
Gene Name
olfactory receptor 10G2
Gene Aliases
-
Chromosome Location
Chr.7: 83533186 - 83534130 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193481.2 NP_001180410.1 - 802 125

View Full Product Details