Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAVIN4
Entrez Gene ID
714379
Gene Name
caveolae associated protein 4
Gene Aliases
MURC
Chromosome Location
Chr.15: 42322187 - 42332801 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001111821.4 XP_001111821.1 - 831 117
GenBank mRNA JU334345.1 - - 482 117

View Full Product Details